accessibility menu, dialog, popup

UserName

HCNO

CAS RN 5799-76-8

IUPAC Name: 4-(prop-2-yn-1-yl)morpholine
Synonyms: 4-(prop-2-yn-1-yl)morpholine
Molecular Weight (g/mol): 125.17
Molecular Formula: C7H11NO
InChi Key: OKDZHAQIKCVKFE-UHFFFAOYSA-N
SMILES: C#CCN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results