missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Propyn-1-yl)morpholine 98.0+%, TCI America™

Catalog No. P2170200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2170200MG 200 mg
P21701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2170200MG Supplier TCI America Supplier No. P2170200MG

Chemical Identifiers

CAS 5799-76-8
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.171
MDL Number MFCD00510919
InChI Key OKDZHAQIKCVKFE-UHFFFAOYSA-N
Synonym 4-Propargylmorpholine
PubChem CID 412941
IUPAC Name 4-prop-2-ynylmorpholine
SMILES C#CCN1CCOCC1

Specifications

Color Yellow
Boiling Point 186°C
Quantity 200 mg
UN Number 1993
Formula Weight 125.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(2-Propyn-1-yl)morpholine