missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Methyl-1H-imidazol-1-yl)benzoic acid hydrochloride hydrate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC61451DA
| Quantity | 1g |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 921938-78-5 | |
| 256.686 | |
| 4-2-methyl-1h-imidazol-1-yl benzoic acid hydrochloride hydrate, 4-2-methylimidazol-1-yl benzoic acid hydrate hydrochloride | |
| 4-(2-methylimidazol-1-yl)benzoic acid;hydrate;hydrochloride |
| C11H13ClN2O3 | |
| PSHJNJCAKINUEP-UHFFFAOYSA-N | |
| 44118781 | |
| CC1=NC=CN1C2=CC=C(C=C2)C(=O)O.O.Cl |
Specifications
| 4-(2-Methyl-1H-imidazol-1-yl)benzoic acid hydrochloride hydrate | |
| 95% | |
| C11H13ClN2O3 | |
| 4-2-methyl-1h-imidazol-1-yl benzoic acid hydrochloride hydrate, 4-2-methylimidazol-1-yl benzoic acid hydrate hydrochloride | |
| CC1=NC=CN1C2=CC=C(C=C2)C(=O)O.O.Cl | |
| 256.686 | |
| 256.69 |
| 921938-78-5 | |
| Amber Glass Bottle | |
| 1g | |
| PSHJNJCAKINUEP-UHFFFAOYSA-N | |
| 4-(2-methylimidazol-1-yl)benzoic acid;hydrate;hydrochloride | |
| 44118781 | |
| 95% |
Safety and Handling
Warning