missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Methyl-1H-imidazol-1-yl)benzoic acid hydrochloride hydrate, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 921938-78-5 |
|---|---|
| Molecular Formula | C11H13ClN2O3 |
| Molecular Weight (g/mol) | 256.686 |
| InChI Key | PSHJNJCAKINUEP-UHFFFAOYSA-N |
| Synonym | 4-2-methyl-1h-imidazol-1-yl benzoic acid hydrochloride hydrate, 4-2-methylimidazol-1-yl benzoic acid hydrate hydrochloride |
| PubChem CID | 44118781 |
| IUPAC Name | 4-(2-methylimidazol-1-yl)benzoic acid;hydrate;hydrochloride |
| SMILES | CC1=NC=CN1C2=CC=C(C=C2)C(=O)O.O.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CC61451CB
|
N/A | 250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC61451DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC61451DE
|
N/A | 5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 921938-78-5 | |
| 256.686 | |
| 4-2-methyl-1h-imidazol-1-yl benzoic acid hydrochloride hydrate, 4-2-methylimidazol-1-yl benzoic acid hydrate hydrochloride | |
| 4-(2-methylimidazol-1-yl)benzoic acid;hydrate;hydrochloride |
| C11H13ClN2O3 | |
| PSHJNJCAKINUEP-UHFFFAOYSA-N | |
| 44118781 | |
| CC1=NC=CN1C2=CC=C(C=C2)C(=O)O.O.Cl |
Specifications
| 921938-78-5 | |
| C11H13ClN2O3 | |
| PSHJNJCAKINUEP-UHFFFAOYSA-N | |
| 4-(2-methylimidazol-1-yl)benzoic acid;hydrate;hydrochloride | |
| 44118781 | |
| 95% |
| 95% | |
| 4-2-methyl-1h-imidazol-1-yl benzoic acid hydrochloride hydrate, 4-2-methylimidazol-1-yl benzoic acid hydrate hydrochloride | |
| CC1=NC=CN1C2=CC=C(C=C2)C(=O)O.O.Cl | |
| 256.686 | |
| 256.69 | |
| 4-(2-Methyl-1H-imidazol-1-yl)benzoic acid hydrochloride hydrate |
Safety and Handling
Warning