missing translation for 'onlineSavingsMsg'
Learn More

[4-(2-Methyl-1,3-thiazol-4-yl)phenyl]methanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 857283-96-6
Molecular Formula C11H11NOS
Molecular Weight (g/mol) 205.275
MDL Number MFCD07772836
InChI Key RJHSPNFHEAFWDW-UHFFFAOYSA-N
Synonym 4-2-methyl-1,3-thiazol-4-yl phenyl methanol, 4-2-methylthiazol-4-yl phenyl methanol, benzenemethanol,4-2-methyl-4-thiazolyl, 4-2-methyl-1,3-thiazol-4-yl phenyl methan-1-ol
PubChem CID 7164608
IUPAC Name [4-(2-methyl-1,3-thiazol-4-yl)phenyl]methanol
SMILES CC1=NC(=CS1)C2=CC=C(C=C2)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC40309CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC40309DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

857283-96-6
205.275
RJHSPNFHEAFWDW-UHFFFAOYSA-N
7164608
CC1=NC(=CS1)C2=CC=C(C=C2)CO
C11H11NOS
MFCD07772836
4-2-methyl-1,3-thiazol-4-yl phenyl methanol, 4-2-methylthiazol-4-yl phenyl methanol, benzenemethanol,4-2-methyl-4-thiazolyl, 4-2-methyl-1,3-thiazol-4-yl phenyl methan-1-ol
[4-(2-methyl-1,3-thiazol-4-yl)phenyl]methanol

Specifications

857283-96-6
100.0
C11H11NOS
4-2-methyl-1,3-thiazol-4-yl phenyl methanol, 4-2-methylthiazol-4-yl phenyl methanol, benzenemethanol,4-2-methyl-4-thiazolyl, 4-2-methyl-1,3-thiazol-4-yl phenyl methan-1-ol
CC1=NC(=CS1)C2=CC=C(C=C2)CO
205.275
205.28
[4-(2-Methyl-1, 3-thiazol-4-yl)phenyl]methanol
96.5
97%
MFCD07772836
RJHSPNFHEAFWDW-UHFFFAOYSA-N
[4-(2-methyl-1,3-thiazol-4-yl)phenyl]methanol
7164608
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents