missing translation for 'onlineSavingsMsg'
Learn More

[4-(2-Methyl-1,3-thiazol-4-yl)phenyl]methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC40309CB

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC40309CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

857283-96-6
205.275
RJHSPNFHEAFWDW-UHFFFAOYSA-N
7164608
CC1=NC(=CS1)C2=CC=C(C=C2)CO
C11H11NOS
MFCD07772836
4-2-methyl-1,3-thiazol-4-yl phenyl methanol, 4-2-methylthiazol-4-yl phenyl methanol, benzenemethanol,4-2-methyl-4-thiazolyl, 4-2-methyl-1,3-thiazol-4-yl phenyl methan-1-ol
[4-(2-methyl-1,3-thiazol-4-yl)phenyl]methanol

Specifications

[4-(2-Methyl-1, 3-thiazol-4-yl)phenyl]methanol
96.5
97%
C11H11NOS
MFCD07772836
RJHSPNFHEAFWDW-UHFFFAOYSA-N
[4-(2-methyl-1,3-thiazol-4-yl)phenyl]methanol
7164608
97%
857283-96-6
100.0
Amber Glass Bottle
250mg
4-2-methyl-1,3-thiazol-4-yl phenyl methanol, 4-2-methylthiazol-4-yl phenyl methanol, benzenemethanol,4-2-methyl-4-thiazolyl, 4-2-methyl-1,3-thiazol-4-yl phenyl methan-1-ol
CC1=NC(=CS1)C2=CC=C(C=C2)CO
205.275
205.28
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

Promotions

Promotions