missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyethyl)phenol 98.0+%, TCI America™

Catalog No. M116725G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M116725G 25 g
M1167500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. M116725G missing translation for 'mfr' TCI America missing translation for 'supplierNo' M116725G

Chemical Identifiers

CAS 56718-71-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00017537
InChI Key FAYGEALAEQKPDI-UHFFFAOYSA-N
Synonym 4-2-methoxyethyl phenol, p-2-methoxyethyl phenol, phenol, 4-2-methoxyethyl, 4-methoxyethylphenol, unii-py1w0akw8i, metoprolol impurity b, metoprolol impurity i, py1w0akw8i, 4-hydroxyphenylethyl methyl ether
PubChem CID 92516
IUPAC Name 4-(2-methoxyethyl)phenol
SMILES COCCC1=CC=C(C=C1)O

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 125°C
Quantity 25 g
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Methoxyethyl)phenol
EINECSNumber (3)-3711
TSCA No