missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Keto-1-benzimidazolinyl)piperidine, 97%
CAS: 20662-53-7 | C12H15N3O | 217.27 g/mol
$125.37 - $457.97
Chemical Identifiers
| CAS | 20662-53-7 |
|---|---|
| Molecular Formula | C12H15N3O |
| Molecular Weight (g/mol) | 217.27 |
| MDL Number | MFCD00005714 |
| InChI Key | BYNBAMHAURJNTR-UHFFFAOYSA-N |
| Synonym | 4-2-keto-1-benzimidazolinyl piperidine, 1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 4-2-keto-1-benzimidozolinyl piperidine, unii-4rp0aed2pc, 1-piperidin-4-yl-1h-benzimidazol-2-ol, 4rp0aed2pc, 1-4-piperidinyl-1,3-dihydro-2h-benzimidazol-2-one, 1-piperidin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-piperidin-4-yl-3h-1,3-benzodiazol-2-one, 1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one |
| PubChem CID | 88638 |
| SMILES | O=C1NC2=CC=CC=C2N1C1CCNCC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC156700010
|
Thermo Scientific Chemicals
156700010 |
1 g | Glass bottle |
Each for $125.37
|
|
||||
|
AC156700050
|
Thermo Scientific Chemicals
156700050 |
5 g | Glass bottle |
Each for $457.97
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 20662-53-7 | |
| 217.27 | |
| BYNBAMHAURJNTR-UHFFFAOYSA-N | |
| 88638 |
| C12H15N3O | |
| MFCD00005714 | |
| 4-2-keto-1-benzimidazolinyl piperidine, 1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 4-2-keto-1-benzimidozolinyl piperidine, unii-4rp0aed2pc, 1-piperidin-4-yl-1h-benzimidazol-2-ol, 4rp0aed2pc, 1-4-piperidinyl-1,3-dihydro-2h-benzimidazol-2-one, 1-piperidin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-piperidin-4-yl-3h-1,3-benzodiazol-2-one, 1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one | |
| O=C1NC2=CC=CC=C2N1C1CCNCC1 |
Specifications
| 20662-53-7 | |
| White to Yellow | |
| 97% | |
| C12H15N3O | |
| 1 g | |
| Solubility in water: 3g/L (20°C). Other solubilities: 32g/100mL methanol- 13g/100mL ethanol,7g/100mL 2-propanol- 0.75g/100mL acetone,3.1g/100mL methylene chloride- 9.4g/100mL n,n-dmf | |
| O=C1NC2=CC=CC=C2N1C1CCNCC1 | |
| 217.27 | |
| 217.27 | |
| Powder |
| 183.0°C to 185.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005714 | |
| 4-2-keto-1-benzimidazolinyl piperidine, 1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 4-2-keto-1-benzimidozolinyl piperidine, unii-4rp0aed2pc, 1-piperidin-4-yl-1h-benzimidazol-2-ol, 4rp0aed2pc, 1-4-piperidinyl-1,3-dihydro-2h-benzimidazol-2-one, 1-piperidin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one, 1-piperidin-4-yl-3h-1,3-benzodiazol-2-one, 1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one | |
| BYNBAMHAURJNTR-UHFFFAOYSA-N | |
| 4-(2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)piperidin-1-ium | |
| 88638 | |
| 97% | |
| 4-(2-Keto-1-benzimidazolinyl)piperidine |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 243-950-3
RUO – Research Use Only