missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)cyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. H12535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12535G 5 g
H12531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12535G Supplier TCI America Supplier No. H12535G

Chemical Identifiers

CAS 74058-21-2
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
InChI Key SZIBVWWQOOVXHS-UHFFFAOYSA-N
Synonym 2-(4-Hydroxycyclohexyl)ethanol, 4-Hydroxycyclohexaneethanol
PubChem CID 13751153
IUPAC Name 4-(2-hydroxyethyl)cyclohexan-1-ol
SMILES C1CC(CCC1CCO)O

Specifications

Color Yellow
Boiling Point 175°C
Quantity 5 g
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(2-Hydroxyethyl)cyclohexanol (cis- and trans- mixture)