accessibility menu, dialog, popup

UserName

CAS RN 74058-21-2

OHOH

CAS RN 74058-21-2

IUPAC Name: 4-(2-hydroxyethyl)cyclohexan-1-ol
Synonyms: 4-(2-hydroxyethyl)cyclohexan-1-ol
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: SZIBVWWQOOVXHS-UHFFFAOYSA-N
SMILES: OCCC1CCC(O)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results