missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)benzonitrile, 98%

Catalog No. p-7051473
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4128303 1 g
AA4128306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4128303 Supplier Thermo Scientific Chemicals Supplier No. 04128303
Only null left

Organic intermediate | CAS: 69395-13-7 | C9H9NO | 147.177 g/mol

4-(2-Hydroxyethyl)benzonitrile is used to produce 4-(2-bromo-ethyl)-benzonitrile. It is used as organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69395-13-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00052461
InChI Key RBSJBNYPTGMZIH-UHFFFAOYSA-N
Synonym 4-2-hydroxyethyl benzonitrile, benzonitrile, 4-2-hydroxyethyl, 2-4-cyanophenyl ethanol, 2-4-cyanophenyl ethan-1-one, 4-2-hydroxyethyl benzenecarbonitrile, 4-cyanobenzeneethanol, p-cyanophenethylalcohol, p-cyanophenethyl alcohol, 4-cyanophenethyl alcohol, acmc-20a4x6
PubChem CID 2800815
IUPAC Name 4-(2-hydroxyethyl)benzonitrile
SMILES C1=CC(=CC=C1CCO)C#N

Specifications

Boiling Point 116°C to 118°C (0.5 mmHg)
Quantity 1 g
UN Number UN3439
Formula Weight 147.18
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-(2-Hydroxyethyl)benzonitrile
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only