CAS RN 69395-13-7
CAS RN 69395-13-7
IUPAC Name:
4-(2-hydroxyethyl)benzonitrile
Synonyms:
4-(2-hydroxyethyl)benzonitrile
Molecular Weight (g/mol):
147.18
Molecular Formula:
C9H9NO
InChi Key:
RBSJBNYPTGMZIH-UHFFFAOYSA-N
SMILES:
OCCC1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
4-(2-Hydroxyethyl)benzonitrile, 98%
CAS: 69395-13-7 Molecular Formula: C9H9NO Molecular Weight (g/mol): 147.177 MDL Number: MFCD00052461 InChI Key: RBSJBNYPTGMZIH-UHFFFAOYSA-N Synonym: 4-2-hydroxyethyl benzonitrile,benzonitrile, 4-2-hydroxyethyl,2-4-cyanophenyl ethanol,2-4-cyanophenyl ethan-1-one,4-2-hydroxyethyl benzenecarbonitrile,4-cyanobenzeneethanol,p-cyanophenethylalcohol,p-cyanophenethyl alcohol,4-cyanophenethyl alcohol,acmc-20a4x6 PubChem CID: 2800815 IUPAC Name: 4-(2-hydroxyethyl)benzonitrile SMILES: C1=CC(=CC=C1CCO)C#N