missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2-Hydroxyethoxy)benzaldehyde 98.0+%, TCI America™
Supplier: TCI America H085925G
Specifications
| 4-(2-Hydroxyethoxy)benzaldehyde | |
| 40°C | |
| MFCD00191450 | |
| VCDGTEZSUNFOKA-UHFFFAOYSA-N | |
| 4-(2-hydroxyethoxy)benzaldehyde | |
| 89178 | |
| ≥98.0% (GC) |
| 22042-73-5 | |
| C9H10O3 | |
| 25 g | |
| OCCOC1=CC=C(C=O)C=C1 | |
| 166.18 | |
| 166.18 | |
| Crystalline Powder |
Chemical Identifiers
| 22042-73-5 | |
| 166.18 | |
| VCDGTEZSUNFOKA-UHFFFAOYSA-N | |
| 4-(2-hydroxyethoxy)benzaldehyde |
| C9H10O3 | |
| MFCD00191450 | |
| 89178 | |
| OCCOC1=CC=C(C=O)C=C1 |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator