missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethoxy)benzaldehyde 98.0+%, TCI America™

Catalog No. p-7130767
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08595G 5 g
H085925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08595G Supplier TCI America Supplier No. H08595G
Only null left

Chemical Identifiers

CAS 22042-73-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00191450
InChI Key VCDGTEZSUNFOKA-UHFFFAOYSA-N
PubChem CID 89178
IUPAC Name 4-(2-hydroxyethoxy)benzaldehyde
SMILES OCCOC1=CC=C(C=O)C=C1

Specifications

Melting Point 40°C
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Hydroxyethoxy)benzaldehyde
TSCA Yes
Recommended Storage Refrigerator