Learn More
4-(2-Cyano-2-propyl)benzeneboronic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H53173MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsBenzene boronic acid mediated the ortho specific hydroxyalkylation of phenols by aldehydes. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. It can be
Specifications
| 4-(2-Cyano-2-propyl)benzeneboronic acid | |
| 146°C to 148°C | |
| MFCD06659832 | |
| 2-4-boronophenyl-2-methylpropanenitrile, 4-2-cyanopropan-2-yl phenylboronic acid, 4-1-cyano-1-methylethyl phenyl boronic acid, 2-4-boronophenyl-2-methylpropionitrile, 4-2-cyanopropan-2-yl benzeneboronic acid, 4-2-cyanopropan-2-yl phenyl boronic acid, 4-1-cyano-1-methylethyl phenylboronic acid, 4-2-cyanoprop-2-yl benzeneboronic acid, boronic acid, 4-1-cyano-1-methylethyl phenyl | |
| BNXBFDTWYHAEIR-UHFFFAOYSA-N | |
| [4-(2-cyanopropan-2-yl)phenyl]boronic acid | |
| 44119324 | |
| 97% |
| 850568-67-1 | |
| C10H12BNO2 | |
| 250 mg | |
| Soluble in water. [9g/100 mL (20°C)] | |
| B(C1=CC=C(C=C1)C(C)(C)C#N)(O)O | |
| 189.021 | |
| 189.02 |
Chemical Identifiers
| 850568-67-1 | |
| 189.021 | |
| BNXBFDTWYHAEIR-UHFFFAOYSA-N | |
| 44119324 | |
| B(C1=CC=C(C=C1)C(C)(C)C#N)(O)O |
| C10H12BNO2 | |
| MFCD06659832 | |
| 2-4-boronophenyl-2-methylpropanenitrile, 4-2-cyanopropan-2-yl phenylboronic acid, 4-1-cyano-1-methylethyl phenyl boronic acid, 2-4-boronophenyl-2-methylpropionitrile, 4-2-cyanopropan-2-yl benzeneboronic acid, 4-2-cyanopropan-2-yl phenyl boronic acid, 4-1-cyano-1-methylethyl phenylboronic acid, 4-2-cyanoprop-2-yl benzeneboronic acid, boronic acid, 4-1-cyano-1-methylethyl phenyl | |
| [4-(2-cyanopropan-2-yl)phenyl]boronic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures