missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloroethyl)morpholine hydrochloride, 99%

Catalog No. aal0577122
Change view
Click to view available options
Quantity:
100 g
500 g
Catalog No. Quantity
AAL0577122 100 g
AAL0577136 500 g
2 options

Catalog No. AAL0577122

Supplier: Thermo Scientific Chemicals L0577122

Only null left

CAS: 3647-69-6 | C6H13Cl2NO | 186.076 g/mol

4-(2-Chloroethyl)morpholine hydrochloride (MOC) is used as intermediate for the synthesis of pharmaceuticals (e.g. floredil, morinamide, nimorazole and pholcodine). It is also used as Agrochemical Intermediates, dyestuff Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Chloroethyl)morpholine hydrochloride (MOC) is used as intermediate for the synthesis of pharmaceuticals (e.g. floredil, morinamide, nimorazole and pholcodine). It is also used as Agrochemical Intermediates, dyestuff Intermediates.

Solubility
Soluble in water, and methanol.

Notes
Hygroscopic. Do not store together with acids. Store under dry inert gas. Protect from humidity and water. Incompatible materials are acids, moisture.

Chemical Identifiers

CAS 3647-69-6
Molecular Formula C6H13Cl2NO
Molecular Weight (g/mol) 186.076
MDL Number MFCD00012797
InChI Key NBJHDLKSWUDGJG-UHFFFAOYSA-N
Synonym 4-2-chloroethyl morpholine hydrochloride, n-2-chloroethyl morpholine hydrochloride, 2-morpholinoethylchloride hydrochloride, morpholine, 4-2-chloroethyl-, hydrochloride, 4-2-chloroethyl morpholine hcl, chloromorpholinoethane hydrochloride, 2-chloroethyl morpholine hydrochloride, 2-morpholinoethyl chloride hydrochloride, 4-2-chloroethyl-morpholine hydrochloride, 4-2-chloroethyl morpholinium chloride
PubChem CID 77210
IUPAC Name 4-(2-chloroethyl)morpholine;hydrochloride
SMILES C1COCCN1CCCl.Cl

Specifications

Melting Point 184°C to 186°C
Quantity 100 g
UN Number UN2923
Beilstein 3684083
Sensitivity Hygroscopic
Solubility Information Soluble in water,and methanol.
Formula Weight 186.08
Percent Purity 99%
Chemical Name or Material 4-(2-Chloroethyl)morpholine hydrochloride
Hazard Category H301-H312-H314-H317-H335
Hazard Statement GHS H Statement
H301-H314-H318-H312-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 222-881-2
RTECSNumber QE0260000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only