missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloroethyl)benzoic acid, 97%

Catalog No. AAL0406818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0406818 50 g
AAL0406809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0406818 Supplier Thermo Scientific Chemicals Supplier No. L0406818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 20849-78-9 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20849-78-9
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00013995
InChI Key OOAPBGPLZAFZSO-UHFFFAOYSA-N
PubChem CID 88712
IUPAC Name 4-(2-chloroethyl)benzoic acid
SMILES OC(=O)C1=CC=C(CCCl)C=C1

Specifications

Melting Point 187°C to 189°C
Quantity 50 g
Beilstein 2084041
Synonym 4-2-chloroethyl benzoic acid, benzoic acid, 4-2-chloroethyl, acmc-1cklh, 4-carboxyphenethyl chloride, p-b-chloroethyl benzoic acid, 4-2-chloro-ethyl benzoic acid, 4-2-chloro-ethyl-benzoic acid, benzoic acid,4-2-chloroethyl, 1-4-carboxyphenyl-2-chloroethane, p-beta-chloroethyl benzoic acid
Formula Weight 184.62
Percent Purity 97%
Chemical Name or Material 4-(2-Chloroethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-080-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only