missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)cyclohexylamine (cis- and trans- mixture) 97.0+%, TCI America™

Catalog No. A26091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26091G 1 g
A26095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26091G Supplier TCI America Supplier No. A26091G

Chemical Identifiers

CAS 202256-86-8
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
InChI Key VFGQLNVWQRKCAU-UHFFFAOYSA-N
Synonym 2-(4-Aminocyclohexyl)ethylamine, 1-Amino-4-(2-aminoethyl)cyclohexane
PubChem CID 21646682
IUPAC Name 4-(2-aminoethyl)cyclohexan-1-amine
SMILES C1CC(CCC1CCN)N

Specifications

Color Colorless
Boiling Point 129°C
Quantity 1 g
UN Number 2735
Formula Weight 142.25
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 4-(2-Aminoethyl)cyclohexylamine (cis- and trans- mixture)