missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)cyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7117814
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22661G 1 g
A22665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22661G Supplier TCI America Supplier No. A22661G

Chemical Identifiers

CAS 148356-06-3
Molecular Formula C8H17NO
Molecular Weight (g/mol) 143.23
MDL Number MFCD11520548
InChI Key FNHBFOVJIPXNFL-UHFFFAOYSA-N
PubChem CID 17773621
IUPAC Name 4-(2-aminoethyl)cyclohexan-1-ol
SMILES NCCC1CCC(O)CC1

Specifications

Color Yellow
Boiling Point 256°C
Quantity 1 g
UN Number 2735
Formula Weight 143.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(2-Aminoethyl)cyclohexanol (cis- and trans- mixture)