accessibility menu, dialog, popup

UserName

NH2OH

CAS RN 148356-06-3

IUPAC Name: 4-(2-aminoethyl)cyclohexan-1-ol
Synonyms: 4-(2-aminoethyl)cyclohexan-1-ol
Molecular Weight (g/mol): 143.23
Molecular Formula: C8H17NO
InChi Key: FNHBFOVJIPXNFL-UHFFFAOYSA-N
SMILES: NCCC1CCC(O)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results