missing translation for 'onlineSavingsMsg'
Learn More

4-(2,4-Dinitrobenzyl)pyridine, TCI America™

Catalog No. D25431G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25431G Supplier TCI America Supplier No. D25431G
Only null left

Chemical Identifiers

CAS 1603-85-6
Molecular Formula C12H9N3O4
Molecular Weight (g/mol) 259.22
MDL Number MFCD01321164
InChI Key ZMNNEUAACIBIEC-UHFFFAOYSA-N
PubChem CID 3953545
IUPAC Name 4-[(2,4-dinitrophenyl)methyl]pyridine
SMILES [O-][N+](=O)C1=CC(=C(CC2=CC=NC=C2)C=C1)[N+]([O-])=O

Specifications

Melting Point 78°C
Color Black-Purple
Quantity 1 g
Formula Weight 259.22
Physical Form Crystalline Powder
Chemical Name or Material 4-(2,4-Dinitrobenzyl)pyridine