CAS RN 1603-85-6
CAS RN 1603-85-6
IUPAC Name:
4-[(2,4-dinitrophenyl)methyl]pyridine
Synonyms:
4-[(2,4-dinitrophenyl)methyl]pyridine
Molecular Weight (g/mol):
259.22
Molecular Formula:
C12H9N3O4
InChi Key:
ZMNNEUAACIBIEC-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(=C(CC2=CC=NC=C2)C=C1)[N+]([O-])=O
1
–
1
of
1
results
1
4-(2,4-Dinitrobenzyl)pyridine, TCI America™
CAS: 1603-85-6 Molecular Formula: C12H9N3O4 Molecular Weight (g/mol): 259.22 MDL Number: MFCD01321164 InChI Key: ZMNNEUAACIBIEC-UHFFFAOYSA-N PubChem CID: 3953545 IUPAC Name: 4-[(2,4-dinitrophenyl)methyl]pyridine SMILES: [O-][N+](=O)C1=CC(=C(CC2=CC=NC=C2)C=C1)[N+]([O-])=O