missing translation for 'onlineSavingsMsg'
Learn More

4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetic acid, 98+%

Catalog No. p-7036563
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5487403 1 g
AAH5487406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5487403 Supplier Thermo Scientific Chemicals Supplier No. H5487403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 126828-35-1 | C32H29NO7 | 539.584 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126828-35-1
Molecular Formula C32H29NO7
Molecular Weight (g/mol) 539.584
MDL Number MFCD00153509
InChI Key UPMGJEMWPQOACJ-UHFFFAOYSA-N
Synonym rink amide linker, 4-2,4-dimethoxyphenyl fmoc-amino methyl phenoxyacetic acid, 2-4-9h-fluoren-9-yl methoxy carbonyl amino 2,4-dimethoxyphenyl methyl phenoxy acetic acid, rink amide linker;, fmoc-knorr, knorr linker;, fmoc-knorr-linker, ambotzrl-1027, pubchem12817, acmc-20amr2
PubChem CID 2761459
IUPAC Name 2-[4-[(2,4-dimethoxyphenyl)-(9H-fluoren-9-ylmethoxycarbonylamino)methyl]phenoxy]acetic acid
SMILES COC1=CC(=C(C=C1)C(C2=CC=C(C=C2)OCC(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)OC

Specifications

Melting Point ∽180°C (decomposition)
Quantity 1 g
Formula Weight 539.58
Percent Purity ≥98%
Chemical Name or Material 4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetic acid
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only