missing translation for 'onlineSavingsMsg'
Learn More

4-[(2,4-Dimethoxyphenyl)[(9H-fluoren-9-ylmethoxy)carbonylamino]methyl]phenoxyacetic Acid 98.0+%, TCI America™

Catalog No. D26725G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26725G Supplier TCI America Supplier No. D26725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 126828-35-1
Molecular Formula C32H29NO7
Molecular Weight (g/mol) 539.584
MDL Number MFCD00153509
InChI Key UPMGJEMWPQOACJ-UHFFFAOYSA-N
Synonym 4-[(2,4-Dimethoxyphenyl)(Fmoc-amino)methyl]phenoxyacetic Acid
PubChem CID 2761459
IUPAC Name 2-[4-[(2,4-dimethoxyphenyl)-(9H-fluoren-9-ylmethoxycarbonylamino)methyl]phenoxy]acetic acid
SMILES COC1=CC(=C(C=C1)C(C2=CC=C(C=C2)OCC(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)OC

Specifications

Melting Point 180°C
Color White
Quantity 5 g
Formula Weight 539.58
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(2,4-Dimethoxyphenyl)[(9H-fluoren-9-ylmethoxy)carbonylamino]methyl]phenoxyacetic Acid
TSCA No
Recommended Storage Refrigerator