missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-([2,2':6',2″-Terpyridin]-4'-yl)benzoic Acid 98.0+%, TCI America™
Supplier: TCI America T34051G
Specifications
| 4-([2,2′:6′,2′′-Terpyridin]-4′-yl)benzoic Acid | |
| 338°C | |
| C22H15N3O2 | |
| 1 g | |
| RPRRPKJKACRJDD-UHFFFAOYSA-N | |
| 4-(2,6-dipyridin-2-ylpyridin-4-yl)benzoic acid | |
| 15903886 | |
| ≥98.0% (GC) |
| 158014-74-5 | |
| White-Yellow | |
| MFCD22056799 | |
| 4′C-(4-Carboxyphenyl)-2,2′C:6′C,2′C′C-terpyridine | |
| C1=CC=NC(=C1)C2=CC(=CC(=N2)C3=CC=CC=N3)C4=CC=C(C=C4)C(=O)O | |
| 353.381 | |
| 353.38 | |
| Crystalline Powder |
Chemical Identifiers
| 158014-74-5 | |
| 353.381 | |
| RPRRPKJKACRJDD-UHFFFAOYSA-N | |
| 15903886 | |
| C1=CC=NC(=C1)C2=CC(=CC(=N2)C3=CC=CC=N3)C4=CC=C(C=C4)C(=O)O |
| C22H15N3O2 | |
| MFCD22056799 | |
| 4′C-(4-Carboxyphenyl)-2,2′C:6′C,2′C′C-terpyridine | |
| 4-(2,6-dipyridin-2-ylpyridin-4-yl)benzoic acid |
Safety and Handling
TSCA : No