missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Tetrazol-5-yl)benzaldehyde, 99%

Catalog No. AAH50470MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50470MD 250 mg
AAH5047003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50470MD Supplier Thermo Scientific Chemicals Supplier No. H50470MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 74815-22-8 | C8H6N4O | 174.163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74815-22-8
Molecular Formula C8H6N4O
Molecular Weight (g/mol) 174.163
MDL Number MFCD01311033
InChI Key SRGPTCYCHZMFOR-UHFFFAOYSA-N
Synonym 4-1h-tetrazol-5-yl benzaldehyde, 4-2h-tetrazol-5-yl benzaldehyde, 4-2h-1,2,3,4-tetrazol-5-yl benzaldehyde, 4-2h-tetrazol-5-yl-benzaldehyde, benzaldehyde, 4-1h-tetrazol-5-yl, 4-1h-1,2,3,4-tetraazol-5-yl benzaldehyde, 4-1h-1,2,3,4-tetrazol-5-yl benzaldehyde, 4-tetrazol-5-yl benzaldehyde, 4-tetrazol-5-yl-benzaldehyde, 4-tetra-zol-5-yl-benzaldehyde
PubChem CID 2760606
IUPAC Name 4-(2H-tetrazol-5-yl)benzaldehyde
SMILES C1=CC(=CC=C1C=O)C2=NNN=N2

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 174.16
Percent Purity 99%
Chemical Name or Material 4-(1H-Tetrazol-5-yl)benzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only