accessibility menu, dialog, popup

UserName

ONNHNN

CAS RN 74815-22-8

IUPAC Name: 4-(2H-1,2,3,4-tetrazol-5-yl)benzaldehyde
Synonyms: 4-(2H-1,2,3,4-tetrazol-5-yl)benzaldehyde
Molecular Weight (g/mol): 174.16
Molecular Formula: C8H6N4O
InChi Key: SRGPTCYCHZMFOR-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C1=NNN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results