missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-ylmethyl)benzylamine hydrochloride , Tech., Thermo Scientific™

Catalog No. p-3485529
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC56063DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC56063DA Supplier Thermo Scientific Chemicals Supplier No. CC56063DA
Only null left

Chemical Identifiers

CAS 904696-62-4
Molecular Formula C11H14ClN3
Molecular Weight (g/mol) 223.70
MDL Number MFCD07690519
InChI Key QUVMWCVEYSYIOW-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-yl methyl phenyl methanamine hydrochloride, 4-1h-pyrazol-1-ylmethyl benzylamine hydrochloride, 4-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 1-4-1h-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 1-4-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 4-pyrazolylmethyl phenyl methylamine, chloride, 4-1h-pyrazol-1-ylmethyl phenyl methanamine hydrochloride, 4-1h-pyrazol-1-yl methyl phenyl methanaminehydrochloride, 1-4-1h-pyrazol-1-yl methyl phenyl methanamine-hydrogen chloride 1/1
PubChem CID 16269819
IUPAC Name [4-(pyrazol-1-ylmethyl)phenyl]methanamine;hydrochloride
SMILES Cl.NCC1=CC=C(CN2C=CC=N2)C=C1

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 223.71
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(1H-pyrazol-1-ylmethyl)benzylamine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.