missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Imidazol-1-ylmethyl)-N-methylbenzylamine 1.5 oxalate 0.5 hydrate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC43396DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC43396DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

910036-84-9
291.307
AIQAEMDKVTYASZ-UHFFFAOYSA-N
24229491
CNCC1=CC=C(C=C1)CN2C=CN=C2.C(=O)(C(=O)O)O
C14H17N3O4
MFCD09064952
oxalic acid; 4-imidazol-1-ylmethyl phenyl methyl methyl amine, oxalic acid-1-4-1h-imidazol-1-yl methyl phenyl-n-methylmethanamine 1/1
1-[4-(imidazol-1-ylmethyl)phenyl]-N-methylmethanamine;oxalic acid

Specifications

4-(1H-imidazol-1-ylmethyl)-N-methylbenzylamine 1.5 oxalate 0.5 hydrate
94.5
95%
C14H17N3O4
MFCD09064952
AIQAEMDKVTYASZ-UHFFFAOYSA-N
1-[4-(imidazol-1-ylmethyl)phenyl]-N-methylmethanamine;oxalic acid
24229491
95%
910036-84-9
100.0
Amber Glass Bottle
1g
oxalic acid; 4-imidazol-1-ylmethyl phenyl methyl methyl amine, oxalic acid-1-4-1h-imidazol-1-yl methyl phenyl-n-methylmethanamine 1/1
CNCC1=CC=C(C=C1)CN2C=CN=C2.C(=O)(C(=O)O)O
291.307
345.34
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions