missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-triazol-1-ylmethyl)aniline, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC45714CB

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC45714CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

119192-10-8
174.207
ZGLQVRIVLWGDNA-UHFFFAOYSA-N
821219
C1=CC(=CC=C1CN2C=NC=N2)N
C9H10N4
MFCD00973306
4-1h-1,2,4-triazol-1-ylmethyl aniline, 4-1h-1,2,4-triazol-1-yl methyl aniline, 1-4-aminobenzyl-1,2,4-triazole, 4-1,2,4-triazol-1-ylmethyl aniline, 4-1h-1,2,4-triazol-1-yl-methyl benzenamine, 4-1,2,4 triazol-1-ylmethylphenylamine, benzenamine, 4-1h-1,2,4-triazol-1-ylmethyl, 4-1,2,4 triazol-1-ylmethyl-phenylamine, 4-1h-1,2,4-triazol-1-yl-methyl aniline, 4-1h-1,2,4-triazol-1-ylmethyl benzenamine
4-(1,2,4-triazol-1-ylmethyl)aniline

Specifications

4-(1H-1, 2, 4-triazol-1-ylmethyl)aniline
96.5
97%
C9H10N4
MFCD00973306
ZGLQVRIVLWGDNA-UHFFFAOYSA-N
4-(1,2,4-triazol-1-ylmethyl)aniline
821219
97%
119192-10-8
100.0
Amber Glass Bottle
250mg
4-1h-1,2,4-triazol-1-ylmethyl aniline, 4-1h-1,2,4-triazol-1-yl methyl aniline, 1-4-aminobenzyl-1,2,4-triazole, 4-1,2,4-triazol-1-ylmethyl aniline, 4-1h-1,2,4-triazol-1-yl-methyl benzenamine, 4-1,2,4 triazol-1-ylmethylphenylamine, benzenamine, 4-1h-1,2,4-triazol-1-ylmethyl, 4-1,2,4 triazol-1-ylmethyl-phenylamine, 4-1h-1,2,4-triazol-1-yl-methyl aniline, 4-1h-1,2,4-triazol-1-ylmethyl benzenamine
C1=CC(=CC=C1CN2C=NC=N2)N
174.207
174.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents