accessibility menu, dialog, popup

UserName

NNNH2N

CAS RN 119192-10-8

IUPAC Name: 4-[(1H-1,2,4-triazol-1-yl)methyl]aniline
Synonyms: 4-(1,2,4-triazol-1-ylmethyl)aniline
Molecular Weight (g/mol): 174.21
Molecular Formula: C9H10N4
InChi Key: ZGLQVRIVLWGDNA-UHFFFAOYSA-N
SMILES: NC1=CC=C(CN2C=NC=N2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)