missing translation for 'onlineSavingsMsg'
Learn More

4-(11-Hydroxyundecyloxy)benzaldehyde 98.0+%, TCI America™

Catalog No. p-7130925
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12161G 1 g
H12165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12161G Supplier TCI America Supplier No. H12161G

Chemical Identifiers

CAS 124389-14-6
Molecular Formula C18H28O3
Molecular Weight (g/mol) 292.42
MDL Number MFCD09038527
InChI Key YUNZXZMRQBKWQP-UHFFFAOYSA-N
PubChem CID 14965977
IUPAC Name 4-[(11-hydroxyundecyl)oxy]benzaldehyde
SMILES OCCCCCCCCCCCOC1=CC=C(C=O)C=C1

Specifications

Melting Point 64°C
Color White-Yellow
Quantity 1 g
Formula Weight 292.42
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(11-Hydroxyundecyloxy)benzaldehyde