accessibility menu, dialog, popup

UserName

OOOH

CAS RN 124389-14-6

IUPAC Name: 4-[(11-hydroxyundecyl)oxy]benzaldehyde
Synonyms: 4-[(11-hydroxyundecyl)oxy]benzaldehyde
Molecular Weight (g/mol): 292.42
Molecular Formula: C18H28O3
InChi Key: YUNZXZMRQBKWQP-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCCOC1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results