missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,10-Diaza-12-crown 4-Ether 96.0+%, TCI America™
$287.06 - $1705.60
Chemical Identifiers
| CAS | 294-92-8 |
|---|---|
| Molecular Formula | C8H20N2O2 |
| Molecular Weight (g/mol) | 176.26 |
| MDL Number | MFCD00142571 |
| InChI Key | PWJHXHMUGFXPSN-UHFFFAOYSA-P |
| Synonym | 4,10-diaza-12-crown 4-ether, 1,7-diaza-12-crown-4, 4,10-dioxa-1,7-diazacyclododecane, diaza-12-crown-4, 4,10-diaza-12-crown4-ether, 1,7-diaza-12-crown |
| PubChem CID | 67522 |
| IUPAC Name | 1,7-dioxa-4,10-diazacyclododecane-4,10-diium |
| SMILES | C1COCC[NH2+]CCOCC[NH2+]1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D2743100MG
|
TCI America
D2743100MG |
100 mg |
Each for $287.06
|
|
|||||
|
D27431G
|
TCI America
D27431G |
1 g |
Each for $1,705.60
|
|
|||||
Chemical Identifiers
| 294-92-8 | |
| 176.26 | |
| PWJHXHMUGFXPSN-UHFFFAOYSA-P | |
| 67522 | |
| C1COCC[NH2+]CCOCC[NH2+]1 |
| C8H20N2O2 | |
| MFCD00142571 | |
| 4,10-diaza-12-crown 4-ether, 1,7-diaza-12-crown-4, 4,10-dioxa-1,7-diazacyclododecane, diaza-12-crown-4, 4,10-diaza-12-crown4-ether, 1,7-diaza-12-crown | |
| 1,7-dioxa-4,10-diazacyclododecane-4,10-diium |
Specifications
| 294-92-8 | |
| White-Yellow | |
| MFCD00142571 | |
| 4,10-diaza-12-crown 4-ether, 1,7-diaza-12-crown-4, 4,10-dioxa-1,7-diazacyclododecane, diaza-12-crown-4, 4,10-diaza-12-crown4-ether, 1,7-diaza-12-crown | |
| C1COCC[NH2+]CCOCC[NH2+]1 | |
| 176.26 | |
| 174.24 | |
| Crystalline Powder |
| 86°C | |
| C8H20N2O2 | |
| 100 mg | |
| PWJHXHMUGFXPSN-UHFFFAOYSA-P | |
| 1,7-dioxa-4,10-diazacyclododecane-4,10-diium | |
| 67522 | |
| ≥96.0% (GC,T) | |
| 4,10-Diaza-12-crown 4-Ether |
Safety and Handling
TSCA : No