accessibility menu, dialog, popup

UserName

CAS RN 294-92-8

ONH2OH2N

CAS RN 294-92-8

IUPAC Name: 1,7-dioxa-4,10-diazacyclododecane-4,10-diium
Synonyms: 1,7-dioxa-4,10-diazacyclododecane-4,10-diium
Molecular Weight (g/mol): 176.26
Molecular Formula: C8H20N2O2
InChi Key: PWJHXHMUGFXPSN-UHFFFAOYSA-P
SMILES: C1COCC[NH2+]CCOCC[NH2+]1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results