missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,10-Diaza-12-crown 4-Ether 96.0+%, TCI America™
Specifications
Specifications
| Melting Point | 86°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 174.24 |
| Percent Purity | ≥96.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,10-Diaza-12-crown 4-Ether |
Safety and Handling
| TSCA | No |