missing translation for 'onlineSavingsMsg'
Learn More

4,10-Diaza-12-crown 4-Ether 96.0+%, TCI America™

Catalog No. D2743100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2743100MG 100 mg
D27431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2743100MG Supplier TCI America Supplier No. D2743100MG
Only null left

Chemical Identifiers

CAS 294-92-8
Molecular Formula C8H20N2O2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00142571
InChI Key PWJHXHMUGFXPSN-UHFFFAOYSA-P
Synonym 4,10-diaza-12-crown 4-ether, 1,7-diaza-12-crown-4, 4,10-dioxa-1,7-diazacyclododecane, diaza-12-crown-4, 4,10-diaza-12-crown4-ether, 1,7-diaza-12-crown
PubChem CID 67522
IUPAC Name 1,7-dioxa-4,10-diazacyclododecane-4,10-diium
SMILES C1COCC[NH2+]CCOCC[NH2+]1

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 100 mg
Formula Weight 174.24
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,10-Diaza-12-crown 4-Ether