missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)pyridine, 98%

CAS: 2456-81-7 | C9H12N2 | 148.209 g/mol

Supplier:  Thermo Scientific Chemicals B2398914

 View more versions of this product

Catalog No. AAB2398914


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-(1-Pyrrolidinyl)pyridine
54°C to 58°C
142°C (287°F)
MFCD00006415
UN2923
4-pyrrolidinopyridine, 4-pyrrolidin-1-yl pyridine, 4-1-pyrrolidinyl pyridine, pyridine, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-pyridine, 4-pyrrolidinylpyridine, 1-4-pyridyl pyrrolidine, zlchem 148, 4-pyrrolidinepyridine, 4-pyrrollidinopyridine
C1CCN(C1)C2=CC=NC=C2
148.209
148.21
2456-81-7
171°C to 173°C (21 mmHg)
C9H12N2
25 g
472443
RGUKYNXWOWSRET-UHFFFAOYSA-N
4-pyrrolidin-1-ylpyridine
75567
98%

Chemical Identifiers

2456-81-7
148.209
RGUKYNXWOWSRET-UHFFFAOYSA-N
75567
C1CCN(C1)C2=CC=NC=C2
C9H12N2
MFCD00006415
4-pyrrolidinopyridine, 4-pyrrolidin-1-yl pyridine, 4-1-pyrrolidinyl pyridine, pyridine, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-pyridine, 4-pyrrolidinylpyridine, 1-4-pyridyl pyrrolidine, zlchem 148, 4-pyrrolidinepyridine, 4-pyrrollidinopyridine
4-pyrrolidin-1-ylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 219-534-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only