missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)pyridine, 98%

Catalog No. AAB2398906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2398906 5 g
AAB2398914 25 g
AAB2398922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2398906 Supplier Thermo Scientific Chemicals Supplier No. B2398906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2456-81-7 | C9H12N2 | 148.209 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2456-81-7
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.209
MDL Number MFCD00006415
InChI Key RGUKYNXWOWSRET-UHFFFAOYSA-N
Synonym 4-pyrrolidinopyridine, 4-pyrrolidin-1-yl pyridine, 4-1-pyrrolidinyl pyridine, pyridine, 4-1-pyrrolidinyl, 4-pyrrolidin-1-yl-pyridine, 4-pyrrolidinylpyridine, 1-4-pyridyl pyrrolidine, zlchem 148, 4-pyrrolidinepyridine, 4-pyrrollidinopyridine
PubChem CID 75567
IUPAC Name 4-pyrrolidin-1-ylpyridine
SMILES C1CCN(C1)C2=CC=NC=C2

Specifications

Melting Point 54°C to 58°C
Boiling Point 171°C to 173°C (21 mmHg)
Flash Point 142°C (287°F)
Quantity 5 g
UN Number UN2923
Beilstein 472443
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 4-(1-Pyrrolidinyl)pyridine
Hazard Category H301-H314
Hazard Statement GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 219-534-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only