missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrenyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. P23051G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23051G Supplier TCI America Supplier No. P23051G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-(1-Pyrenyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 872050-52-7
Molecular Formula C22H15BO2
Quantity 1 g
MDL Number MFCD28130386
Synonym 4-(1-Pyrenyl)benzeneboronic Acid
InChI Key ISMRPUXCQLIMJL-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(C=C1)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
IUPAC Name [4-(pyren-1-yl)phenyl]boronic acid
Molecular Weight (g/mol) 322.17
PubChem CID 59217173
Formula Weight 322.17
Physical Form Crystalline Powder
Show More Show Less