missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1-Piperazinyl)benzonitrile 98.0+%, TCI America™
$244.19 - $730.71
Chemical Identifiers
| CAS | 68104-63-2 |
|---|---|
| Molecular Formula | C11H13N3 |
| Molecular Weight (g/mol) | 187.246 |
| MDL Number | MFCD01320890 |
| InChI Key | DJJNYEXRPRQXPD-UHFFFAOYSA-N |
| Synonym | 4-piperazin-1-yl benzonitrile, 1-4-cyanophenyl piperazine, 4-piperazinobenzonitrile, 4-1-piperazinyl benzonitrile, 1-4-cyanophenyl-piperazine, 4-piperazin-1-yl-benzonitrile, 4-piperazinobenzenecarbonitrile, benzonitrile, 4-1-piperazinyl, 4-piperazinylbenzenecarbonitrile, 4-cyanophenylpiperazine |
| PubChem CID | 2733995 |
| IUPAC Name | 4-piperazin-1-ylbenzonitrile |
| SMILES | C1CN(CCN1)C2=CC=C(C=C2)C#N |
Chemical Identifiers
| 68104-63-2 | |
| 187.246 | |
| DJJNYEXRPRQXPD-UHFFFAOYSA-N | |
| 2733995 | |
| C1CN(CCN1)C2=CC=C(C=C2)C#N |
| C11H13N3 | |
| MFCD01320890 | |
| 4-piperazin-1-yl benzonitrile, 1-4-cyanophenyl piperazine, 4-piperazinobenzonitrile, 4-1-piperazinyl benzonitrile, 1-4-cyanophenyl-piperazine, 4-piperazin-1-yl-benzonitrile, 4-piperazinobenzenecarbonitrile, benzonitrile, 4-1-piperazinyl, 4-piperazinylbenzenecarbonitrile, 4-cyanophenylpiperazine | |
| 4-piperazin-1-ylbenzonitrile |
Specifications
| 68104-63-2 | |
| White-Yellow | |
| MFCD01320890 | |
| 4-piperazin-1-yl benzonitrile, 1-4-cyanophenyl piperazine, 4-piperazinobenzonitrile, 4-1-piperazinyl benzonitrile, 1-4-cyanophenyl-piperazine, 4-piperazin-1-yl-benzonitrile, 4-piperazinobenzenecarbonitrile, benzonitrile, 4-1-piperazinyl, 4-piperazinylbenzenecarbonitrile, 4-cyanophenylpiperazine | |
| C1CN(CCN1)C2=CC=C(C=C2)C#N | |
| 187.246 | |
| 187.25 | |
| Crystalline Powder |
| 83°C | |
| C11H13N3 | |
| 1 g | |
| DJJNYEXRPRQXPD-UHFFFAOYSA-N | |
| 4-piperazin-1-ylbenzonitrile | |
| 2733995 | |
| ≥98.0% (GC,T) | |
| 4-(1-Piperazinyl)benzonitrile |
Safety and Handling
TSCA : No