missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Piperazinyl)benzonitrile 98.0+%, TCI America™

Catalog No. P24491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24491G 1 g
P24495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24491G Supplier TCI America Supplier No. P24491G
Only null left

Chemical Identifiers

CAS 68104-63-2
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.246
MDL Number MFCD01320890
InChI Key DJJNYEXRPRQXPD-UHFFFAOYSA-N
Synonym 4-piperazin-1-yl benzonitrile, 1-4-cyanophenyl piperazine, 4-piperazinobenzonitrile, 4-1-piperazinyl benzonitrile, 1-4-cyanophenyl-piperazine, 4-piperazin-1-yl-benzonitrile, 4-piperazinobenzenecarbonitrile, benzonitrile, 4-1-piperazinyl, 4-piperazinylbenzenecarbonitrile, 4-cyanophenylpiperazine
PubChem CID 2733995
IUPAC Name 4-piperazin-1-ylbenzonitrile
SMILES C1CN(CCN1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 1 g
Formula Weight 187.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1-Piperazinyl)benzonitrile