missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Naphthylvinyl)pyridine 98.0+%, TCI America™

Catalog No. N04915G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N04915G Supplier TCI America Supplier No. N04915G

Chemical Identifiers

CAS 16375-56-7
Molecular Formula C17H13N
Molecular Weight (g/mol) 231.298
MDL Number MFCD00059136
InChI Key FLKDRTOVVLNOLV-CMDGGOBGSA-N
Synonym 1-(4-Pyridylvinyl)naphthalene
PubChem CID 5475238
IUPAC Name 4-[(E)-2-naphthalen-1-ylethenyl]pyridine
SMILES C1=CC=C2C(=C1)C=CC=C2C=CC3=CC=NC=C3

Specifications

Melting Point 82°C
Quantity 5 g
Formula Weight 231.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1-Naphthylvinyl)pyridine