missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Imidazolyl)phenol, 97%

Catalog No. aaa1321103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1321103 1 g
AAA1321106 5 g
AAA1321114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1321103 Supplier Thermo Scientific Chemicals Supplier No. A1321103
Only null left

CAS: 10041-02-8 | C9H8N2O | 160.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10041-02-8
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD00005281
InChI Key CYKCUAPYWQDIKR-UHFFFAOYSA-N
Synonym 4-imidazol-1-yl phenol, 4-1h-imidazol-1-yl phenol, 4-imidazol-1-yl-phenol, 4-1-imidazolyl phenol, 1-p-hydroxyphenyl imidazole, phenol, 4-1h-imidazol-1-yl, 1-4-hydroxyphenyl imidazole, 4-imidazolylphenol, 4‐ 1h‐imidazol‐1‐yl phenol compound 1, p-1-imidazolyl phenol
PubChem CID 82315
IUPAC Name 4-imidazol-1-ylphenol
SMILES OC1=CC=C(C=C1)N1C=CN=C1

Specifications

Melting Point 204°C to 208°C
Quantity 1 g
Beilstein 509999
Formula Weight 160.18
Percent Purity 97%
Chemical Name or Material 4-(1-Imidazolyl)phenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-121-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only