accessibility menu, dialog, popup

UserName

CAS RN 10041-02-8

IUPAC Name: 4-(1H-imidazol-1-yl)phenol
Synonyms: 4-(1H-imidazol-1-yl)phenol
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: CYKCUAPYWQDIKR-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)N1C=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results