missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Thiazol-2-yl)benzaldehyde, 95%, Thermo Scientific™

Chemical Identifiers

CAS 198904-53-9
Molecular Formula C10H7NOS
Molecular Weight (g/mol) 189.23
MDL Number MFCD06797780
InChI Key LQLBILPEELCFQI-UHFFFAOYSA-N
Synonym 4-thiazol-2-yl-benzaldehyde, 4-1,3-thiazol-2-yl benzaldehyde, 4-thiazol-2-yl benzaldehyde, benzaldehyde,4-2-thiazolyl, benzaldehyde, 4-2-thiazolyl, 4-2-thiazolyl benzaldehyde, 4-thiazol-2-ylbenzaldehyde, 4-2-thiazolyl-benzaldehyde
PubChem CID 10535655
IUPAC Name 4-(1,3-thiazol-2-yl)benzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=NC=CS1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC40204CB
View Documents Promotion Details
N/A 250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC40204DA
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC40204DE
View Documents Promotion Details
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

198904-53-9
189.23
LQLBILPEELCFQI-UHFFFAOYSA-N
10535655
O=CC1=CC=C(C=C1)C1=NC=CS1
C10H7NOS
MFCD06797780
4-thiazol-2-yl-benzaldehyde, 4-1,3-thiazol-2-yl benzaldehyde, 4-thiazol-2-yl benzaldehyde, benzaldehyde,4-2-thiazolyl, benzaldehyde, 4-2-thiazolyl, 4-2-thiazolyl benzaldehyde, 4-thiazol-2-ylbenzaldehyde, 4-2-thiazolyl-benzaldehyde
4-(1,3-thiazol-2-yl)benzaldehyde

Specifications

198904-53-9
C10H7NOS
4-thiazol-2-yl-benzaldehyde, 4-1,3-thiazol-2-yl benzaldehyde, 4-thiazol-2-yl benzaldehyde, benzaldehyde,4-2-thiazolyl, benzaldehyde, 4-2-thiazolyl, 4-2-thiazolyl benzaldehyde, 4-thiazol-2-ylbenzaldehyde, 4-2-thiazolyl-benzaldehyde
O=CC1=CC=C(C=C1)C1=NC=CS1
189.23
189.24
4-(1, 3-Thiazol-2-yl)benzaldehyde
95%
MFCD06797780
LQLBILPEELCFQI-UHFFFAOYSA-N
4-(1,3-thiazol-2-yl)benzaldehyde
10535655
95%
Videos
SDS
Documents

Documents