missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1,3-Thiazol-2-yl)benzaldehyde, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC40204CB
| Quantity | 250mg |
|---|
Chemical Identifiers
| 198904-53-9 | |
| 189.23 | |
| LQLBILPEELCFQI-UHFFFAOYSA-N | |
| 10535655 | |
| O=CC1=CC=C(C=C1)C1=NC=CS1 |
| C10H7NOS | |
| MFCD06797780 | |
| 4-thiazol-2-yl-benzaldehyde, 4-1,3-thiazol-2-yl benzaldehyde, 4-thiazol-2-yl benzaldehyde, benzaldehyde,4-2-thiazolyl, benzaldehyde, 4-2-thiazolyl, 4-2-thiazolyl benzaldehyde, 4-thiazol-2-ylbenzaldehyde, 4-2-thiazolyl-benzaldehyde | |
| 4-(1,3-thiazol-2-yl)benzaldehyde |
Specifications
| 4-(1, 3-Thiazol-2-yl)benzaldehyde | |
| 95% | |
| MFCD06797780 | |
| 4-thiazol-2-yl-benzaldehyde, 4-1,3-thiazol-2-yl benzaldehyde, 4-thiazol-2-yl benzaldehyde, benzaldehyde,4-2-thiazolyl, benzaldehyde, 4-2-thiazolyl, 4-2-thiazolyl benzaldehyde, 4-thiazol-2-ylbenzaldehyde, 4-2-thiazolyl-benzaldehyde | |
| O=CC1=CC=C(C=C1)C1=NC=CS1 | |
| 189.23 | |
| 189.24 |
| 198904-53-9 | |
| C10H7NOS | |
| 250mg | |
| LQLBILPEELCFQI-UHFFFAOYSA-N | |
| 4-(1,3-thiazol-2-yl)benzaldehyde | |
| 10535655 | |
| 95% |