missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Dioxolan-2-yl)benzonitrile, 97%, Thermo Scientific™

$464.27 - $464.27

Chemical Identifiers

CAS 66739-89-7
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00052570
InChI Key AQXQCWAXQJVTKU-UHFFFAOYSA-N
PubChem CID 2778428
IUPAC Name 4-(1,3-dioxolan-2-yl)benzonitrile
SMILES N#CC1=CC=C(C=C1)C1OCCO1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5896304
View Documents
Thermo Scientific Chemicals
H5896304
2 g
Each for $464.27
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

66739-89-7
175.19
AQXQCWAXQJVTKU-UHFFFAOYSA-N
4-(1,3-dioxolan-2-yl)benzonitrile
C10H9NO2
MFCD00052570
2778428
N#CC1=CC=C(C=C1)C1OCCO1

Specifications

66739-89-7
MFCD00052570
UN3439
AQXQCWAXQJVTKU-UHFFFAOYSA-N
4-(1,3-dioxolan-2-yl)benzonitrile
2778428
97%
C10H9NO2
2 g
4-1,3-dioxolan-2-yl benzonitrile, 4-1,3 dioxolan-2-yl-benzonitrile, 4-1,3 dioxolane-2-yl-benzonitrile, 4-1,3-dioxolane-2-yl-benzonitrile, benzonitrile,4-1,3-dioxolan-2-yl, benzonitrile, 4-1,3-dioxolan-2-yl, 4-1,3-dioxolan-2-yl benzenecarbonitrile
N#CC1=CC=C(C=C1)C1OCCO1
175.19
175.19
4-(1,3-Dioxolan-2-yl)benzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents