accessibility menu, dialog, popup

UserName

OON

CAS RN 66739-89-7

IUPAC Name: 4-(1,3-dioxolan-2-yl)benzonitrile
Synonyms: 4-(1,3-dioxolan-2-yl)benzonitrile
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: AQXQCWAXQJVTKU-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)C1OCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results