missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Dioxolan-2-yl)benzonitrile, 97%, Thermo Scientific™

Catalog No. AAH5896304
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5896304 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5896304 Supplier Thermo Scientific Chemicals Supplier No. H5896304
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 66739-89-7
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00052570
InChI Key AQXQCWAXQJVTKU-UHFFFAOYSA-N
PubChem CID 2778428
IUPAC Name 4-(1,3-dioxolan-2-yl)benzonitrile
SMILES N#CC1=CC=C(C=C1)C1OCCO1

Specifications

Quantity 2 g
UN Number UN3439
Synonym 4-1,3-dioxolan-2-yl benzonitrile, 4-1,3 dioxolan-2-yl-benzonitrile, 4-1,3 dioxolane-2-yl-benzonitrile, 4-1,3-dioxolane-2-yl-benzonitrile, benzonitrile,4-1,3-dioxolan-2-yl, benzonitrile, 4-1,3-dioxolan-2-yl, 4-1,3-dioxolan-2-yl benzenecarbonitrile
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 4-(1,3-Dioxolan-2-yl)benzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures